Biphenyl-4,4'-dicarboxylic acid
Leverantör: Apollo Scientific
Varning
Synonyms: 4,4'-Biphenyldicarboxylic acid, [1,1'-Biphenyl]-4,4'-dicarboxylic acid
OR2318-25G OR2318-5G
APOSOR2318-25GEA 465 SEK
APOSOR2318-25G APOSOR2318-5G
Biphenyl-4,4'-dicarboxylic acid
Biphenyl-4,4'-dicarboxylic acid
Formel: C₁₄H₁₀O₄
MW: 242,23 g/mol
Smältpunkt: >300 °C
Storage Temperature: Omgivande
MDL Number: MFCD00002554
CAS nummer: 787-70-2
EINECS: 212-328-3