Pyrrole-2-carboxylic acid 97%

Leverantör: Apollo Scientific
Varning

OR23111-5G
APOSOR23111-5GEA 720 SEK
APOSOR23111-5G
Pyrrole-2-carboxylic acid 97%
Pyrrole-2-carboxylic acid
Formel: C₅H₅NO₂
MW: 111.10 g/mol
Smältpunkt: 204…209 °C (subl.)
Storage Temperature: Omgivande
MDL Number: MFCD00005219
CAS nummer: 634-97-9
EINECS: 211-221-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR