1-Cyclohexenylboronic acid pinacol ester 95%
Leverantör: Apollo Scientific
Varning
Synonyms: 1-Cyclohexeneboronic acid pinacol ester, 2-(Cyclohex-1-en-1-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 1-Cyclohexene-1-boronic acid pinacol ester
OR2302-1G OR2302-5G
APOSOR2302-1GEA 380 SEK
APOSOR2302-1G APOSOR2302-5G
1-Cyclohexenylboronic acid pinacol ester 95%
1-Cyclohexenylboronic acid pinacol ester
Formel: C₁₂H₂₁BO₂
MW: 208,1049 g/mol
Kokpunkt: 125 °C/0,23mmHg
Densitet: 0,968 g/cm³ (25 °C)
Flampunkt: >100 °C
Storage Temperature: Omgivande
MDL Number: MFCD05663845
CAS nummer: 141091-37-4