5-Methyl-3-phenylisoxazole-4-carboxylic acid
Leverantör: Apollo Scientific
Varning
Synonyms: Oxacillin Related Compound C
OR22978-5G OR22978-25G
APOSOR22978-5GEA 295 SEK
APOSOR22978-5G APOSOR22978-25G
5-Methyl-3-phenylisoxazole-4-carboxylic acid
5-Methyl-3-phenylisoxazole-4-carboxylic acid
Formel: C₁₁H₉NO₃
MW: 203,197 g/mol
Smältpunkt: 191…194 °C
Storage Temperature: Omgivande
MDL Number: MFCD00003153
CAS nummer: 1136-45-4
EINECS: 214-497-9