N-Decyl-N,N-dimethyl-3-ammonio-1-propanesulphonate(sulphobetaine-10, SB-10) 98%
Leverantör: Apollo Scientific
Synonyms: 3-[Dec-1-yl(dimethyl)ammonio]propane-1-sulphonate, 3-(Decyldimethylazaniumyl)propane-1-sulphonate
OR2270T-25G OR2270T-5G
APOSOR2270T-25GEA 2725 SEK
APOSOR2270T-25G APOSOR2270T-5G
N-Decyl-N,N-dimethyl-3-ammonio-1-propanesulphonate(sulphobetaine-10, SB-10) 98%
N-Decyl-N,N-dimethyl-3-ammonio-1-propanesulphonate(sulphobetaine-10, SB-10)
A zwitterionic detergent.
Formel: C₁₅H₃₃NO₃S
MW: 307,4924 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00036908
CAS nummer: 15163-36-7