(S)-2-Aminopropanamide hydrochloride
Leverantör: Apollo Scientific
Varning
Synonyms: H-Ala-NH2.HCl, (S)-2-Aminopropionamide Hydrochloride, L-Alanamine Hydrochloride, H-Ala-NH₂·HCl, L-Alaninamide hydrochloride
OR22615-100G OR22615-25G
APOSOR22615-100GEA 1125 SEK
APOSOR22615-100G APOSOR22615-25G
(S)-2-Aminopropanamide hydrochloride
(S)-2-Aminopropanamide hydrochloride
Formel: C₃H₉ClN₂O
MW: 124.57 g/mol
Smältpunkt: 212…217 °C
Storage Temperature: Omgivande
MDL Number: MFCD00066145
CAS nummer: 33208-99-0