3-Hydroxy-2-(hydroxymethyl)pyridine hydrochloride 95%
Leverantör: Apollo Scientific
Varning
Synonyms: 2-Pyridinemethanol, 3-hydroxy-, hydrochloride, 2-(hydroxymethyl)pyridin-3-ol, chloride, 3-Hydroxy-2-pyridinemethanol hydrochloride, 3-Hydroxy-2-(hydroxymethyl)pyridine hydrochloride
OR2259-1G OR2259-5G
APOSOR2259-1GEA 175 SEK
APOSOR2259-1G APOSOR2259-5G
3-Hydroxy-2-(hydroxymethyl)pyridine hydrochloride 95%
3-Hydroxy-2-(hydroxymethyl)pyridine hydrochloride
Formel: C₆H₈ClNO₂
MW: 161,585 g/mol
Smältpunkt: 217 °C
Storage Temperature: Omgivande
MDL Number: MFCD00006349
CAS nummer: 14173-30-9
EINECS: 238-021-4