3-(N-Boc-Methylaminomethyl)azetidine hydrochloride
Leverantör: Apollo Scientific
Varning
Synonyms: 3-[(Methylamino)methyl]azetidine hydrochloride, 3-BOC protected
OR2247-500MG OR2247-250MG
APOSOR2247-500MGEA 3200 SEK
APOSOR2247-500MG APOSOR2247-250MG
3-(N-Boc-Methylaminomethyl)azetidine hydrochloride
3-(N-Boc-Methylaminomethyl)azetidine hydrochloride
Formel: C₁₀H₂₁ClN₂O₂
MW: 236.74 g/mol
Storage Temperature: Kylskåp
MDL Number: MFCD09998721
CAS nummer: 1170019-97-2