4-Propoxybenzonitrile

Leverantör: Apollo Scientific
Varning

OR21919-1G OR21919-100MG OR21919-5G
APOSOR21919-1GEA 685 SEK
APOSOR21919-1G APOSOR21919-100MG APOSOR21919-5G
4-Propoxybenzonitrile
4-Propoxybenzonitrile
Formel: C₁₀H₁₁NO
MW: 161.200 g/mol
Smältpunkt: 41 °C
Storage Temperature: Omgivande
MDL Number: MFCD00173874
CAS nummer: 60758-84-1
UN: 3439
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR