(6-Chloroimidazo[1,2-b]pyridazin-3-yl)(phenyl)methanone
Leverantör: Apollo Scientific
Varning
OR21487-1G OR21487-50MG OR21487-250MG OR21487-500MG
APOSOR21487-1GEA 5675 SEK
APOSOR21487-1G APOSOR21487-50MG APOSOR21487-250MG APOSOR21487-500MG
(6-Chloroimidazo[1,2-b]pyridazin-3-yl)(phenyl)methanone
(6-Chloroimidazo[1,2-b]pyridazin-3-yl)(phenyl)methanone
Formel: C₁₃H₈ClN₃O
Storage Temperature: Omgivande
MDL Number: MFCD00108116
CAS nummer: 90734-72-8