N-(2,4-Dimethoxyphenyl)-2-chloroacetamide
Leverantör: Apollo Scientific
Varning
Synonyms: N-(Chloroacetyl)-2,4-dimethoxyaniline
OR21448-1G OR21448-5G
APOSOR21448-1GEA 685 SEK
APOSOR21448-1G APOSOR21448-5G
N-(2,4-Dimethoxyphenyl)-2-chloroacetamide
N-(2,4-Dimethoxyphenyl)-2-chloroacetamide
Formel: C₁₀H₁₂ClNO₃
Storage Temperature: Omgivande
CAS nummer: 101908-41-2