5-Methylanthranil acid
Leverantör: Apollo Scientific
Varning
Synonyms: 2-Amino-5-methylbenzoic acid, 6-Amino-m-toluic acid
OR21443-100G OR21443-25G OR21443-500G OR21443-1KG
APOSOR21443-100GEA 600 SEK
APOSOR21443-100G APOSOR21443-25G APOSOR21443-500G APOSOR21443-1KG
5-Methylanthranil acid
5-Methylanthranil acid
Formel: C₈H₉NO₂
MW: 151,16 g/mol
Smältpunkt: 175 °C (dec.)
Storage Temperature: Omgivande
MDL Number: MFCD00007909
CAS nummer: 2941-78-8
EINECS: 220-932-3