4-Chloroanthranilic acid 98%
Leverantör: Apollo Scientific
Varning
Synonyms: 2-Amino-4-chlorobenzoic acid
OR21298-100G OR21298-500G OR21298-1KG
APOSOR21298-100GEA 585 SEK
APOSOR21298-100G APOSOR21298-500G APOSOR21298-1KG
4-Chloroanthranilic acid 98%
4-Chloroanthranilic acid
Formel: C₇H₆ClNO₂
MW: 171,58 g/mol
Smältpunkt: 231…233 °C (dec.)
Storage Temperature: Omgivande
MDL Number: MFCD00007778
CAS nummer: 89-77-0
EINECS: 201-938-5