3,4-Diaminoanisole
Leverantör: Apollo Scientific
|
Varning
|
Synonyms:
4-Methoxy-o-phenylenediamine
OR21113-1G
OR21113-25G
OR21113-5G
APOSOR21113-1GEA
260
SEK
APOSOR21113-1G
APOSOR21113-25G
APOSOR21113-5G
3,4-Diaminoanisole
3,4-Diaminoanisole
Formel:
C₇H₁₀N₂O Smältpunkt: 46…50 °C Storage Temperature: Kylskåp |
MDL Number:
MFCD00047837 CAS nummer: 102-51-2 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...