Terephthalonitrile 95%

Leverantör: Apollo Scientific
Varning

Synonyms: 1,4-Benzenedicarbonitrile, Terephthalodinitrile, Benzene-1,4-dicarbonitrile, 1,4-Dicyanobenzene

OR19552-100G OR19552-500G
APOSOR19552-100GEA 775 SEK
APOSOR19552-100G APOSOR19552-500G
Terephthalonitrile 95%
Terephthalonitrile
Formel: C₈H₄N₂
MW: 128,13 g/mol
Smältpunkt: 221…225 °C
Storage Temperature: Omgivande
MDL Number: MFCD00001810
CAS nummer: 623-26-7
EINECS: 210-783-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR