2-Aminoisonicotinic acid

Leverantör: Apollo Scientific
Varning

Synonyms: 2-Amino-4-pyridinecarboxylic acid, 2-Aminopyridine-4-carboxylic acid

OR1916-25G OR1916-5G
APOSOR1916-25GEA 550 SEK
APOSOR1916-25G APOSOR1916-5G
2-Aminoisonicotinic acid
2-Aminoisonicotinic acid
Formel: C₆H₆N₂O₂
MW: 138,13 g/mol
Smältpunkt: 300…302 °C
Storage Temperature: Omgivande
MDL Number: MFCD00233714
CAS nummer: 13362-28-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR