4-Methylcinnamic acid
Leverantör: Apollo Scientific
Varning
Synonyms: 3-(p-Tolyl)acrylic acid
OR18348-100G OR18348-25G OR18348-5G
APOSOR18348-100GEA 790 SEK
APOSOR18348-100G APOSOR18348-25G APOSOR18348-5G
4-Methylcinnamic acid
4-Methylcinnamic acid
Formel: C₁₀H₁₀O₂
MW: 162,19 g/mol
Smältpunkt: 196…198 °C
Storage Temperature: Omgivande
MDL Number: MFCD00002697
CAS nummer: 1866-39-3