α,α,α,3,4-Pentachlorotoluene 98%
Leverantör: Apollo Scientific
|
Fara
|
Synonyms:
3,4-Dichlorobenzotrichloride, 1,2-Dichloro-4-(trichloromethyl)benzene
OR17904-25G
OR17904-500G
APOSOR17904-25GEA
1050
SEK
APOSOR17904-25G
APOSOR17904-500G
α,α,α,3,4-Pentachlorotoluene 98%
α,α,α,3,4-Pentachlorotoluene
Formel:
C₇H₃Cl₅ MW: 264,3637 g/mol Smältpunkt: 26 °C Densitet: 1,576 g/cm³ (25 °C) Flampunkt: 110 °C Storage Temperature: Omgivande |
MDL Number:
MFCD00018820 CAS nummer: 13014-24-9 EINECS: 235-869-7 UN: 1759 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...