Monomethyl-trans-1,4-cyclohexanedicarboxylate
Leverantör: Apollo Scientific
Varning
Synonyms: Methyl Hydrogen trans-1,4-Cyclohexanedicarboxylate, trans-4-(Methoxycarbonyl)cyclohexanecarboxylic acid, trans-1,4-Cyclohexanedicarboxylic Acid Monomethyl Ester
OR17609-100G OR17609-25G OR17609-5G
APOSOR17609-100GEA 1400 SEK
APOSOR17609-100G APOSOR17609-25G APOSOR17609-5G
Monomethyl-trans-1,4-cyclohexanedicarboxylate
Monomethyl-trans-1,4-cyclohexanedicarboxylate
Formel: C₉H₁₄O₄
MW: 186,21 g/mol
Smältpunkt: 128…131 °C
Storage Temperature: Omgivande
MDL Number: MFCD01311247
CAS nummer: 15177-67-0