3-Amino-1,2,4-triazine 97%
Leverantör: Apollo Scientific
|
Varning
|
Synonyms:
1,2,4-Triazine-3-ylamine, 1,2,4-Triazin-3-amine, 1,2,4-Triazine-3-amine
OR17565-10G
OR17565-25G
APOSOR17565-10GEA
825
SEK
APOSOR17565-10G
APOSOR17565-25G
3-Amino-1,2,4-triazine 97%
3-Amino-1,2,4-triazine
Formel:
C₃H₄N₄ MW: 96,09 g/mol Smältpunkt: 174…177 °C Storage Temperature: Omgivande |
MDL Number:
MFCD00006459 CAS nummer: 1120-99-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...