2-Amino-2-metylpropanol 95%
Leverantör: Apollo Scientific
Varning
Synonyms: 2-Amino-2-methyl-1-propanol, Isobutanolamine, AMP
OR1740T-1KG OR1740T-500G
APOSOR1740T-1KGEA 1250 SEK
APOSOR1740T-1KG APOSOR1740T-500G
2-Amino-2-metylpropanol 95%
2-Amino-2-metylpropanol
Used for the preparation of buffer solutions, suitable for the determination of alkaline phosphatase.
Formel: (CH₃)₂C(NH₂)CH₂OH
MW: 89,14 g/mol
Kokpunkt: 164…167 °C
Smältpunkt: 24…32 °C
Densitet: 0,934 g/cm³ (25 °C)
Flampunkt: 68 °C
Storage Temperature: Omgivande
MDL Number: MFCD00008051
CAS nummer: 124-68-5
EINECS: 204-709-8