4-Amino-3-hydroxybenzenesulphonic acid 95+%

Leverantör: Apollo Scientific
Varning

OR17109-1G OR17109-500MG
APOSOR17109-1GEA 7725 SEK
APOSOR17109-1G APOSOR17109-500MG
4-Amino-3-hydroxybenzenesulphonic acid 95+%
4-Amino-3-hydroxybenzenesulphonic acid
Formel: C₆H₇NO₄S
Storage Temperature: Kylskåp
MDL Number: MFCD14560533
CAS nummer: 2592-14-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR