6,8-Dibromoimidazo[1,2-a]pyrazine
Leverantör: Apollo Scientific
Varning
OR16939-100G OR16939-25G OR16939-5G
APOSOR16939-100GEA 5075 SEK
APOSOR16939-100G APOSOR16939-25G APOSOR16939-5G
6,8-Dibromoimidazo[1,2-a]pyrazine
6,8-Dibromoimidazo[1,2-a]pyrazine
Formel: C₆H₃Br₂N₃
Storage Temperature: Omgivande
MDL Number: MFCD08460056
CAS nummer: 63744-22-9