2-Iodobenzenesulphonic acid 98%
Leverantör: Apollo Scientific
|
Fara
|
OR16595-1G
OR16595-5G
OR16595-25G
APOSOR16595-1GEA
345
SEK
APOSOR16595-1G
APOSOR16595-5G
APOSOR16595-25G
2-Iodobenzenesulphonic acid 98%
2-Iodobenzenesulphonic acid
Formel:
C₆H₅IO₃S Storage Temperature: Omgivande |
MDL Number:
MFCD01860232 CAS nummer: 63059-25-6 UN: 3261 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...