Tetrahydrofuran-3-carbonyl chloride

Leverantör: Apollo Scientific
Fara

OR16566-1G OR16566-250MG OR16566-100MG
APOSOR16566-1GEA 3550 SEK
APOSOR16566-1G APOSOR16566-250MG APOSOR16566-100MG
Tetrahydrofuran-3-carbonyl chloride
Tetrahydrofuran-3-carbonyl chloride
Formel: C₅H₇ClO₂
MW: 134.56 g/mol
Kokpunkt: 75…76 °C/15mmHg
Storage Temperature: Omgivande
MDL Number: MFCD06656696
CAS nummer: 69595-02-4
UN: 3265
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR