6-Bromo-2(3H)-benzoxazolone 98%

Leverantör: Apollo Scientific
Varning

Synonyms: 6-Bromo-2-benzoxazolinone, 6-Bromo-2-benzoxazolone, 6-Bromo-2-hydroxybenzoxazole, 6-Bromobenzo[d]oxazol-2(3H)-one

OR16545-10G OR16545-5G
APOSOR16545-10GEA 1100 SEK
APOSOR16545-10G APOSOR16545-5G
6-Bromo-2(3H)-benzoxazolone 98%
6-Bromo-2(3H)-benzoxazolone
Formel: C₇H₄BrNO₂
MW: 214,018 g/mol
Smältpunkt: 192…196 °C
Storage Temperature: Omgivande
MDL Number: MFCD00694727
CAS nummer: 19932-85-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR