Sodium octyl sulphate 99%
Leverantör: Apollo Scientific
Varning
Synonyms: Octyl sulphate, sodium salt, Octyl sulphuric acid salt
OR16472-25G OR16472-5G
APOSOR16472-25GEA 4650 SEK
APOSOR16472-25G APOSOR16472-5G
Sodium octyl sulphate 99%
Sodium octyl sulphate
Formel: C₈H₁₇NaO₄S
MW: 232,28 g/mol
Smältpunkt: 210 °C
Storage Temperature: Omgivande
MDL Number: MFCD00007470
CAS nummer: 142-31-4