3-Indoleacrylic acid 98%
Leverantör: Apollo Scientific
|
Varning
|
Synonyms:
IAA, 3-(1-H-Indol-3-yl)-2-propenoic acid, 3-(1H-Indol-3-yl)acrylic acid, 3-(3-Indolyl)acrylic Acid
OR16454-1G
OR16454-5G
APOSOR16454-1GEA
1475
SEK
APOSOR16454-1G
APOSOR16454-5G
3-Indoleacrylic acid 98%
3-Indoleacrylic acid
Formel:
C₁₁H₉NO₂ MW: 187,20 g/mol Smältpunkt: 183…185 °C Storage Temperature: Omgivande |
MDL Number:
MFCD00005633 CAS nummer: 1204-06-4 EINECS: 214-872-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...