3-Iodobenzyl alcohol, Technical Grade
Leverantör: Apollo Scientific
Varning
Synonyms: (3-Iodophenyl)methanol, 3-Iodobenzylalcohol
OR16142-100G OR16142-25G OR16142-5G
APOSOR16142-100GEA 3600 SEK
APOSOR16142-100G APOSOR16142-25G APOSOR16142-5G
3-Iodobenzyl alcohol, Technical Grade
3-Iodobenzyl alcohol
Formel: C₇H₇IO
MW: 234,04 g/mol
Kokpunkt: 254…255 °C
Densitet: 1,842 g/cm³ (25 °C)
Flampunkt: >110 °C
Storage Temperature: Omgivande
MDL Number: MFCD00004635
CAS nummer: 57455-06-8
EINECS: 260-744-9