1-Hydroxy-1-cyclopropanecarboxylic acid 98%
Leverantör: Apollo Scientific
Varning
Synonyms: 1-hydroxycyclopropane-1-carboxylic acid, 1-Hydroxycyclopropanecarboxylic acid
OR15989-1G OR15989-5G OR15989-25G
APOSOR15989-1GEA 260 SEK
APOSOR15989-1G APOSOR15989-5G APOSOR15989-25G
1-Hydroxy-1-cyclopropanecarboxylic acid 98%
1-Hydroxy-1-cyclopropanecarboxylic acid
Formel: C₄H₆O₃
MW: 102.089 g/mol
Smältpunkt: 108…110 °C
Storage Temperature: Omgivande
MDL Number: MFCD00010797
CAS nummer: 17994-25-1