2-Thiazolecarboxylic acid 95%
Leverantör: Apollo Scientific
Varning
Synonyms: Thiazole-2-carboxylic acid
OR15912-5G OR15912-10G
APOSOR15912-5GEA 790 SEK
APOSOR15912-5G APOSOR15912-10G
2-Thiazolecarboxylic acid 95%
2-Thiazolecarboxylic acid
Formel: C₄H₃NO₂S
MW: 129,14 g/mol
Smältpunkt: 91,5…93 °C
Storage Temperature: Omgivande
MDL Number: MFCD02094169
CAS nummer: 14190-59-1