4-Methoxy-N-methylbenzylamine hydrochloride 95%

Leverantör: Apollo Scientific
Varning

OR1574-1G OR1574-5G
APOSOR1574-1GEA 685 SEK
APOSOR1574-1G APOSOR1574-5G
4-Methoxy-N-methylbenzylamine hydrochloride 95%
4-Methoxy-N-methylbenzylamine hydrochloride
Formel: C₉H₁₄ClNO
MW: 187,6700 g/mol
Kokpunkt: 261,2 °C
Storage Temperature: Omgivande
MDL Number: MFCD04117881
CAS nummer: 876-32-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR