(R)-1,4-Diazabicyclo[4.3.0]nonane, Technical Grade
Leverantör: Apollo Scientific
Fara
OR15658-1G OR15658-5G OR15658-10G
APOSOR15658-1GEA 1175 SEK
APOSOR15658-1G APOSOR15658-5G APOSOR15658-10G
(R)-1,4-Diazabicyclo[4.3.0]nonane, Technical Grade
(R)-1,4-Diazabicyclo[4.3.0]nonane
Formel: C₇H₁₄N₂
MW: 126,20 g/mol
Kokpunkt: 65…69 °C/3mmHg
Storage Temperature: Omgivande
MDL Number: MFCD09752820
CAS nummer: 96193-27-0
UN: 3267
ADR: 8,III