3-Bromo-5-(hydroxymethyl)isoxazole
Leverantör: Apollo Scientific
Varning
Synonyms: (3-Bromoisoxazol-5-yl)methanol
OR15590-1G OR15590-250MG
APOSOR15590-1GEA 1200 SEK
APOSOR15590-1G APOSOR15590-250MG
3-Bromo-5-(hydroxymethyl)isoxazole
3-Bromo-5-(hydroxymethyl)isoxazole
Formel: C₄H₄BrNO₂
MW: 177.98 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD04035586
CAS nummer: 25742-00-1