2-Amino-5-chloro-4-methylthiazole 95%
Leverantör: Apollo Scientific
Varning
OR15552-1G OR15552-5G
APOSOR15552-1GEA 1300 SEK
APOSOR15552-1G APOSOR15552-5G
2-Amino-5-chloro-4-methylthiazole 95%
2-Amino-5-chloro-4-methylthiazole
Formel: C₄H₅ClN₂S
MW: 148,61 g/mol
Storage Temperature: Kylskåp
MDL Number: MFCD10699130
CAS nummer: 5316-76-7