1-Pyrimidin-2-ylpiperidin-4-one

Leverantör: Apollo Scientific
Varning

OR15509-1G OR15509-5G
APOSOR15509-1GEA 1200 SEK
APOSOR15509-1G APOSOR15509-5G
1-Pyrimidin-2-ylpiperidin-4-one
1-Pyrimidin-2-ylpiperidin-4-one
Formel: C₉H₁₁N₃O
Smältpunkt: 73…75 °C
Storage Temperature: Omgivande
CAS nummer: 116247-92-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR