3-(Benzoylsulphanyl)-2-methylpropanoic acid

Leverantör: Apollo Scientific
Varning

Synonyms: 3-(Benzoylthio)-2-methylpropanoic acid

OR15471-1G OR15471-5G
APOSOR15471-1GEA 600 SEK
APOSOR15471-1G APOSOR15471-5G
3-(Benzoylsulphanyl)-2-methylpropanoic acid
3-(Benzoylsulphanyl)-2-methylpropanoic acid
Formel: C₁₁H₁₂O₃S
MW: 224,28 g/mol
Smältpunkt: 63…64 °C
Storage Temperature: Omgivande
MDL Number: MFCD09864827
CAS nummer: 74431-50-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR