4-[(1-Oxo-1,3-dihydro-2H-isoindol-2-yl)methyl]-benzenecarboxylic acid
Leverantör: Apollo Scientific
Varning
Synonyms: 4-[(1,3-Dihydro-1-oxo-2H-isoindol-2-yl)methyl]benzoic acid
OR15427-10G OR15427-1G OR15427-5G
APOSOR15427-10GEA 9575 SEK
APOSOR15427-10G APOSOR15427-1G APOSOR15427-5G
4-[(1-Oxo-1,3-dihydro-2H-isoindol-2-yl)methyl]-benzenecarboxylic acid
4-[(1-Oxo-1,3-dihydro-2H-isoindol-2-yl)methyl]benzenecarboxylic acid
Formel: C₁₆H₁₃NO₃
MW: 267.29 g/mol
Smältpunkt: 228…230 °C
Storage Temperature: Omgivande
MDL Number: MFCD06200936
CAS nummer: 503039-50-7