4-Bromo-2-piperidinobenzenecarbaldehyde
Leverantör: Apollo Scientific
Varning
OR15273-1G OR15273-500MG
APOSOR15273-1GEA 1825 SEK
APOSOR15273-1G APOSOR15273-500MG
4-Bromo-2-piperidinobenzenecarbaldehyde
4-Bromo-2-piperidinobenzenecarbaldehyde
Formel: C₁₂H₁₄BrNO
Storage Temperature: Omgivande
MDL Number: MFCD08276958
CAS nummer: 643094-36-4