1-(2-Amino-3-pyridinyl)-1-ethanone

Leverantör: Apollo Scientific
Varning

Synonyms: 1-(2-Aminopyridin-3-yl)ethanone, 3-Acetyl-2-aminopyridine

OR15249-1G OR15249-5G
APOSOR15249-1GEA 840 SEK
APOSOR15249-1G APOSOR15249-5G
1-(2-Amino-3-pyridinyl)-1-ethanone
1-(2-Amino-3-pyridinyl)-1-ethanone
Formel: C₇H₈N₂O
MW: 136,15 g/mol
Smältpunkt: 139…140 °C
Storage Temperature: Omgivande
MDL Number: MFCD02258895
CAS nummer: 65326-33-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR