2,6-Bis[(dimethylamino)methyl]-4-(1-methylbutyl)benzenol
Leverantör: Apollo Scientific
Varning
Synonyms: 2,6-Bis[(dimethylamino)methyl]-4-pent-2-ylphenol
OR15180-10G OR15180-1G OR15180-5G
APOSOR15180-10GEA 10625 SEK
APOSOR15180-10G APOSOR15180-1G APOSOR15180-5G
2,6-Bis[(dimethylamino)methyl]-4-(1-methylbutyl)benzenol
2,6-Bis[(dimethylamino)methyl]-4-(1-methylbutyl)benzenol
Formel: C₁₇H₃₀N₂O
MW: 278,44 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD08443975
CAS nummer: 924868-91-7