1-(4-Amino-3-methoxyphenyl)-1-ethanone 97%

Leverantör: Apollo Scientific
Varning

Synonyms: 4'-Amino-3'-methoxyacetophenone, 1-(4-Amino-3-methoxyphenyl)ethanone

OR15131-1G OR15131-500MG
APOSOR15131-1GEA 5000 SEK
APOSOR15131-1G APOSOR15131-500MG
1-(4-Amino-3-methoxyphenyl)-1-ethanone 97%
1-(4-Amino-3-methoxyphenyl)-1-ethanone
Formel: C₉H₁₁NO₂
MW: 165,19 g/mol
Smältpunkt: 90…92 °C
Storage Temperature: Omgivande
MDL Number: MFCD08443961
CAS nummer: 22106-40-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR