Potassium-4-iodobenzenesulphonate
Leverantör: Apollo Scientific
|
Varning
|
OR15122-1G
OR15122-5G
APOSOR15122-1GEA
1475
SEK
APOSOR15122-1G
APOSOR15122-5G
Potassium-4-iodobenzenesulphonate
Potassium-4-iodobenzenesulphonate
Formel:
C₆H₄IKO₃S Storage Temperature: Omgivande |
MDL Number:
MFCD06656232 CAS nummer: 13035-63-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...