2-Hydrazino-1,3-benzothiazole-6-carbohydrazide
Leverantör: Apollo Scientific
OR15096-1G OR15096-5G
APOSOR15096-1GEA 6325 SEK
APOSOR15096-1G APOSOR15096-5G
2-Hydrazino-1,3-benzothiazole-6-carbohydrazide
2-Hydrazino-1,3-benzothiazole-6-carbohydrazide
Formel: C₈H₉N₅OS
MW: 223.26 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD08444017
CAS nummer: 924869-07-8