5-Aminolevulinic acid hydrochloride 97%
Leverantör: Apollo Scientific
Varning
Synonyms: 5-Amino-4-ketovaleric acid hydrochloride, 5-Amino-4-oxopentanoic acid hydrochloride, δ-Aminolevulinic Acid Hydrochloride
OR1490-1G OR1490-5G OR1490-25G
APOSOR1490-1GEA 260 SEK
APOSOR1490-1G APOSOR1490-5G APOSOR1490-25G
5-Aminolevulinic acid hydrochloride 97%
5-Aminolevulinic acid hydrochloride
Formel: C₅H₁₀ClNO₃
MW: 167.59 g/mol
Smältpunkt: 150 °C (dec.)
Storage Temperature: Frys
MDL Number: MFCD00012869
CAS nummer: 5451-09-2
EINECS: 226-679-5