(6-Methyl-3-pyridinyl)methanol
Leverantör: Apollo Scientific
Varning
Synonyms: 6-Methyl-3-pyridinemethanol, 5-Hydroxymethyl-2-methylpyridine, (6-Methylpyridin-3-yl)methanol, 5-Hydroxymethyl-2-picoline
OR14843-250MG OR14843-5G OR14843-25G
APOSOR14843-250MGEA 600 SEK
APOSOR14843-250MG APOSOR14843-5G APOSOR14843-25G
(6-Methyl-3-pyridinyl)methanol
(6-Methyl-3-pyridinyl)methanol
Formel: C₇H₉NO
MW: 123,16 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD08692003
CAS nummer: 34107-46-5