L-1-Naphthylalanine 95%

Leverantör: Apollo Scientific
Varning

Synonyms: H-1-Nal-OH, 3-(1-Naphthyl)-L-alanine

OR14711-100MG OR14711-5G OR14711-10G
APOSOR14711-100MGEA 175 SEK
APOSOR14711-100MG APOSOR14711-5G APOSOR14711-10G
L-1-Naphthylalanine 95%
L-1-Naphthylalanine
Formel: C₁₃H₁₃NO₂
MW: 215,252 g/mol
Smältpunkt: 254 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00066086
CAS nummer: 55516-54-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR