1-Bromo-4-butylbenzene
Leverantör: Apollo Scientific
|
Varning
|
Synonyms:
(4-Bromophenyl)butane
OR1467-100G
OR1467-250G
OR1467-25G
APOSOR1467-100GEA
755
SEK
APOSOR1467-100G
APOSOR1467-250G
APOSOR1467-25G
1-Bromo-4-butylbenzene
1-Bromo-4-butylbenzene
Formel:
C₁₀H₁₃Br MW: 213,12 g/mol Kokpunkt: 242 °C Densitet: 1,208 g/cm³ (25 °C) Flampunkt: 108 °C Storage Temperature: Omgivande |
MDL Number:
MFCD00040934 CAS nummer: 41492-05-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...