Fmoc-3-(1-naphthyl)-L-alanine 98%
Leverantör: Apollo Scientific
Varning
Synonyms: Fmoc-1-Nal-OH
OR14596-10G OR14596-5G
APOSOR14596-10GEA 2450 SEK
APOSOR14596-10G APOSOR14596-5G
Fmoc-3-(1-naphthyl)-L-alanine 98%
Fmoc-3-(1-naphthyl)-L-alanine
Formel: C₂₈H₂₃NO₄
MW: 437.495 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00151914
CAS nummer: 96402-49-2