Pyridine-2-carboxamidoxime
Leverantör: Apollo Scientific
Varning
Synonyms: 2-Pyridylamidoxime, N'-Hydroxypicolinimidamide, 2-Pyridinecarboxamide Oxime, N-Hydroxypicolinimidamide
OR14504-25G OR14504-5G
APOSOR14504-25GEA 740 SEK
APOSOR14504-25G APOSOR14504-5G
Pyridine-2-carboxamidoxime
Pyridine-2-carboxamidoxime
Formel: C₆H₇N₃O
MW: 137,14 g/mol
Smältpunkt: 120…124 °C
Storage Temperature: Omgivande
MDL Number: MFCD00085159
CAS nummer: 1772-01-6